Gene Transcripts
- Gene Symbol
- Slc17a9
- Entrez Gene ID
- 228993
- Gene Name
- solute carrier family 17, member 9
- Gene Aliases
- 1700019H03Rik, BC019537, Vnut
- Chromosome Location
- Chr. 2: 180725213 - 180742280 on Build GRCm38
- UniGene ID
- Mm.87189
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_183161.3 |
NP_898984.3
|
8, 9 |
954 |
|
GenBank mRNA |
BE850231.1 |
- |
1, 2 |
83 |
|
View Full Product Details