Gene Transcripts

Gene Symbol
Nccrp1
Entrez Gene ID
233038
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
1190020J12Rik, Gm163, NCCRP-1
Chromosome Location
Chr. 7: 28543596 - 28547254 on Build GRCm38
UniGene ID
Mm.224815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081115.1 NP_001074584.1 2, 3 577
GenBank mRNA AK028022.1 - 2, 3 227

View Full Product Details