Gene Transcripts

Gene Symbol
Dmxl2
Entrez Gene ID
235380
Gene Name
Dmx-like 2
Gene Aliases
6330586A12, 6430411K14Rik, E130119P06Rik, mKIAA0856
Chromosome Location
Chr. 9: 54365158 - 54501697 on Build GRCm38
UniGene ID
Mm.93636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172771.2 NP_766359.2 39 8600
GenBank mRNA AK173043.1 - 21 4681
BC059265.1 - 7 593

View Full Product Details