Gene Transcripts

Gene Symbol
Gpatch3
Entrez Gene ID
242691
Gene Name
G patch domain containing 3
Gene Aliases
D930035B09Rik, Gpatc3
Chromosome Location
Chr. 4: 133574745 - 133584242 on Build GRCm38
UniGene ID
Mm.40110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172876.2 NP_766464.1 7 1531
GenBank mRNA AK053051.1 - 7 1532

View Full Product Details