Gene Transcripts
- Gene Symbol
- Rbbp7
- Entrez Gene ID
- 245688
- Gene Name
- retinoblastoma binding protein 7
- Gene Aliases
- AA409861, AI173248, AU019541, BB114024, mRbAp46
- Chromosome Location
- Chr. X: 162760372 - 162779090 on Build GRCm38
- UniGene ID
- Mm.270186
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009031.3 |
NP_033057.3
|
10, 11 |
1408 |
|
GenBank mRNA |
AK076016.1 |
- |
10, 11 |
1333 |
|
AK078349.1 |
- |
10, 11 |
1321 |
|
AK082892.1 |
- |
10, 11 |
1320 |
|
AK135779.1 |
- |
10, 11 |
1322 |
|
AK135869.1 |
- |
9, 10 |
1017 |
|
AK135875.1 |
- |
9, 10 |
1017 |
|
AK135956.1 |
- |
10, 11 |
1322 |
|
AK136110.1 |
- |
10, 11 |
1320 |
|
AK145531.1 |
- |
10, 11 |
1319 |
|
AK145651.1 |
- |
10, 11 |
1302 |
|
AK146014.1 |
- |
10, 11 |
1320 |
|
AK146439.1 |
- |
10, 11 |
1379 |
|
AK146904.1 |
- |
10, 11 |
1321 |
|
AK146967.1 |
- |
10, 11 |
1320 |
|
AK147062.1 |
- |
10, 11 |
1379 |
|
AK148852.1 |
- |
10, 11 |
1348 |
|
AK153913.1 |
- |
10, 11 |
1365 |
|
AK160023.1 |
- |
10, 11 |
1320 |
|
AK214411.1 |
- |
2, 3 |
71 |
|
AK214866.1 |
- |
2, 3 |
71 |
|
AK215897.1 |
- |
2, 3 |
71 |
|
AK219296.1 |
- |
2, 3 |
71 |
|
BC003785.1 |
- |
10, 11 |
1337 |
3600013 |
U35142.1 |
- |
10, 11 |
1347 |
|
View Full Product Details