Gene Transcripts

Gene Symbol
Cntrl
Entrez Gene ID
26920
Gene Name
centriolin
Gene Aliases
6720467O09Rik, AA968343, Cep1, Cep110, IB3/5, Ma2a8, b2b1468Clo
Chromosome Location
Chr. 2: 35109492 - 35178822 on Build GRCm38
UniGene ID
Mm.231332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012018.2 NP_036148.2 33, 34 5603
GenBank mRNA AK131139.1 - 11, 12 1957
BC079536.1 - 21, 22 3628
BC158010.1 - 32, 33 5476 9007318

View Full Product Details