Gene Transcripts

Gene Symbol
Elmod1
Entrez Gene ID
270162
Gene Name
ELMO/CED-12 domain containing 1
Gene Aliases
4831417L10, AI428506, AI834991
Chromosome Location
Chr. 9: 53911457 - 53975310 on Build GRCm38
UniGene ID
Mm.259791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177769.4 NP_808437.2 10 1071
GenBank mRNA AK029207.1 - 10 952
AK132239.1 - 10 1072
AK164094.1 - 223
BC049160.1 - 8 731
BC120566.1 - 9 787
BC144961.1 - 9 787
BC144963.1 - 9 749 9053511

View Full Product Details