Gene Transcripts

Gene Symbol
Smyd4
Entrez Gene ID
319822
Gene Name
SET and MYND domain containing 4
Gene Aliases
G430029E23Rik
Chromosome Location
Chr. 11: 75348433 - 75405707 on Build GRCm38
UniGene ID
Mm.340377
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102611.1 NP_001096081.1 8, 9 2344
GenBank mRNA AK030380.1 - 8, 9 2181
AK089959.1 - 8, 9 2280
AK154507.1 - 7, 8 2030
AK173306.1 - 6, 7 1766
BC049810.1 - 8, 9 2195
BC095952.1 - 8, 9 2241 5037842
BC130220.1 - 5, 6 1681 40141368

View Full Product Details