Gene Transcripts

Gene Symbol
Ptrh1
Entrez Gene ID
329384
Gene Name
peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae)
Gene Aliases
2210013M04Rik, AW046744
Chromosome Location
Chr. 2: 32775738 - 32777593 on Build GRCm38
UniGene ID
Mm.268037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178595.3 NP_848710.1 4 426
GenBank mRNA AK075833.1 - 4 427
BC098218.1 - 4 466 3466952

View Full Product Details