Gene Transcripts

Gene Symbol
Trim67
Entrez Gene ID
330863
Gene Name
tripartite motif-containing 67
Gene Aliases
D130049O21Rik, TNL
Chromosome Location
Chr. 8: 124793019 - 124834704 on Build GRCm38
UniGene ID
Mm.422769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198632.2 NP_941034.2 1 1608
GenBank mRNA AK051455.1 - 1 1610
AK220282.1 - 1 1055
BC094596.1 - 1 1535 5705515

View Full Product Details