Gene Transcripts

Gene Symbol
Cngb1
Entrez Gene ID
333329
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
Cngb1b, Gm1959
Chromosome Location
Chr. 8: 95239043 - 95306585 on Build GRCm38
UniGene ID
Mm.484049
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195413.1 NP_001182342.1 21 2347
GenBank mRNA BC045114.1 - 7 909
HQ116386.1 - 21 2347

View Full Product Details