Gene Transcripts

Gene Symbol
Mtg2
Entrez Gene ID
52856
Gene Name
mitochondrial ribosome associated GTPase 2
Gene Aliases
1810011P19Rik, 2900056P18Rik, D2Bwg0647e, Gtpbp5
Chromosome Location
Chr. 2: 180070593 - 180085902 on Build GRCm38
UniGene ID
Mm.219658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083328.1 NP_001076797.1 7 720
NM_001083329.1 NP_001076798.1 6 626
NM_028422.1 NP_082698.1 7 765
NM_181424.3 NP_852089.2 8 859
GenBank mRNA AK040689.1 - 7 1249
BC025144.1 - 8 858 4483063
BC028465.1 - 2 161
BC030918.1 - 2 905
BC039784.1 - 2 916
DT929445.1 - 4 343

View Full Product Details