Gene Transcripts
- Gene Symbol
- Naa10
- Entrez Gene ID
- 56292
- Gene Name
- N(alpha)-acetyltransferase 10, NatA catalytic subunit
- Gene Aliases
- 2310039H09Rik, Ard1, Ard1a, Te2
- Chromosome Location
- Chr. X: 73916870 - 73921944 on Build GRCm38
- UniGene ID
- Mm.246654
- Species
- Mus musculus
- Gene Symbol
- Arhgap4
- Entrez Gene ID
- 171207
- Gene Name
- Rho GTPase activating protein 4
- Gene Aliases
- A130009C12Rik, A530015A20Rik, Rgc1, c1, mKIAA0131
- Chromosome Location
- Chr. X: 73894352 - 73911346 on Build GRCm38
- UniGene ID
- Mm.482337
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK129060.1 |
- |
|
213 |
|
View Full Product Details