Gene Transcripts

Gene Symbol
Vmn2r93
Entrez Gene ID
627132
Gene Name
vomeronasal 2, receptor 93
Gene Aliases
EG627132
Chromosome Location
Chr. 17: 18298281 - 18326441 on Build GRCm38
UniGene ID
Mm.469862
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104542.1 NP_001098012.1 3 1168

View Full Product Details