Gene Transcripts

Gene Symbol
Vmn2r12
Entrez Gene ID
627569
Gene Name
vomeronasal 2, receptor 12
Gene Aliases
EG627569, Gm6769
Chromosome Location
Chr. 5: 109085849 - 109097864 on Build GRCm38
UniGene ID
Mm.469884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104623.1 NP_001098093.1 3 1133

View Full Product Details