Gene Transcripts

Gene Symbol
Skint8
Entrez Gene ID
639774
Gene Name
selection and upkeep of intraepithelial T cells 8
Gene Aliases
OTTMUSG00000009475
Chromosome Location
Chr. 4: 111919446 - 111950361 on Build GRCm38
UniGene ID
Mm.248185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100466.1 NP_001093936.2 7 1022
GenBank mRNA EU099307.1 - 7 1022

View Full Product Details