Gene Transcripts

Gene Symbol
Sdf2l1
Entrez Gene ID
64136
Gene Name
stromal cell-derived factor 2-like 1
Gene Aliases
Chromosome Location
Chr. 16: 17130138 - 17132383 on Build GRCm38
UniGene ID
Mm.30222
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022324.3 NP_071719.1 3 636
GenBank mRNA AB043006.1 - 3 576
BC023290.1 - 3 590
BC053425.1 - 3 618 6508268
BC053438.1 - 3 618

View Full Product Details