Gene Transcripts

Gene Symbol
Bod1l
Entrez Gene ID
665775
Gene Name
biorientation of chromosomes in cell division 1-like
Gene Aliases
A230054D04Rik, AI853319, FAM44A, mKIAA1327
Chromosome Location
Chr. 5: 41787538 - 41844365 on Build GRCm38
UniGene ID
Mm.158726
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081422.2 NP_001074891.1 10 6223
GenBank mRNA AF013969.1 - 1 1682
AK122495.1 - 1 1367

View Full Product Details