Gene Transcripts

Gene Symbol
H60b
Entrez Gene ID
667281
Gene Name
histocompatibility 60b
Gene Aliases
EG667281
Chromosome Location
Chr. 10: 22273474 - 22289144 on Build GRCm38
UniGene ID
Mm.484254
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177775.1 NP_001171246.1 4 557
GenBank mRNA AB284505.1 - 634

View Full Product Details