Gene Transcripts

Gene Symbol
Higd2a
Entrez Gene ID
67044
Gene Name
HIG1 domain family, member 2A
Gene Aliases
2010110M21Rik, AA690237
Chromosome Location
Chr. 13: 54590231 - 54591147 on Build GRCm38
UniGene ID
Mm.29353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025933.3 NP_080209.1 1 174
GenBank mRNA AK004272.1 - 1 14
AK005269.1 - 1 175
AK008387.1 - 1 171
BC021471.1 - 1 171 5343356

View Full Product Details