Gene Transcripts

Gene Symbol
Cntd1
Entrez Gene ID
68107
Gene Name
cyclin N-terminal domain containing 1
Gene Aliases
1700051C09Rik, 4921513J16Rik, 5430417M23Rik, Cntd
Chromosome Location
Chr. 11: 101279203 - 101288701 on Build GRCm38
UniGene ID
Mm.262369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026562.2 NP_080838.1 1, 2 271
GenBank mRNA AK007245.1 - 1, 2 272
AK049116.1 - 1, 2 379
BC006866.1 - 1, 2 330 3595113

View Full Product Details