Gene Transcripts

Gene Symbol
Fam212a
Entrez Gene ID
68176
Gene Name
family with sequence similarity 212, member A
Gene Aliases
6230427J02Rik, Inka1
Chromosome Location
Chr. 9: 107984223 - 107985916 on Build GRCm38
UniGene ID
Mm.76166
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026597.3 NP_080873.2 2 928
GenBank mRNA AK020090.1 - 2 892
AK076092.1 - 2 892
BC051538.1 - 1 669
BC115538.1 - 2 867 40101503
BQ176112.1 - 1 146

View Full Product Details