Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_197982.3 | NP_932099.2 | 9 | 1235 | |
GenBank mRNA | AK011285.1 | - | 8 | 1074 | |
AK012002.1 | - | 9 | 1142 | ||
AK016960.1 | - | 9 | 1144 | ||
AK088894.1 | - | 3 | 537 | ||
AK137114.1 | - | 1141 | |||
AK145927.1 | - | 9 | 1178 | ||
AK146294.1 | - | 9 | 1180 | ||
AK149475.1 | - | 9 | 1577 | ||
AK150997.1 | - | 9 | 1164 | ||
AK151395.1 | - | 10 | 1204 | ||
AK152394.1 | - | 9 | 1145 | ||
AK162752.1 | - | 1703 | |||
AK164289.1 | - | 9 | 1105 | ||
AK167890.1 | - | 9 | 1105 | ||
AK168079.1 | - | 9 | 1104 | ||
AK172448.1 | - | 9 | 1164 | ||
AK195258.1 | - | 3 | 225 | ||
BC020134.1 | - | 9 | 1123 | 3969048 |