Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026823.2 | NP_081099.1 | 4, 5 | 519 | |
GenBank mRNA | AB185207.1 | - | 4, 5 | 375 | |
AK004080.1 | - | 1, 2 | 47 | ||
BC004035.1 | - | 3, 4 | 250 | ||
BC018479.1 | - | 4, 5 | 519 | 3661329 |