Gene Transcripts
- Gene Symbol
- Odam
- Entrez Gene ID
- 69592
- Gene Name
- odontogenic, ameloblast asssociated
- Gene Aliases
- 2310011G06Rik, APIN
- Chromosome Location
- Chr. 5: 87885025 - 87894174 on Build GRCm38
- UniGene ID
- Mm.389652
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027128.2 |
NP_081404.2
|
3, 4 |
160 |
|
GenBank mRNA |
EF113909.1 |
- |
3, 4 |
160 |
|
View Full Product Details