Gene Transcripts

Gene Symbol
Mrgbp
Entrez Gene ID
73247
Gene Name
MRG/MORF4L binding protein
Gene Aliases
1600027N09Rik, AW060503, C80444
Chromosome Location
Chr. 2: 180581222 - 180586304 on Build GRCm38
UniGene ID
Mm.426558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028479.1 NP_082755.1 6 991
GenBank mRNA AK038431.1 - 4 618
BC042508.1 - 3 1797
BC054431.1 - 4 887
BC085320.1 - 2 2290
BC110993.1 - 5 597 5363496
BC116732.1 - 6 991 40126051
BC116734.1 - 6 991 40126053

View Full Product Details