Gene Transcripts
- Gene Symbol
- Ddit4l
- Entrez Gene ID
- 73284
- Gene Name
- DNA-damage-inducible transcript 4-like
- Gene Aliases
- 1700037B15Rik, 1700108M02Rik, REDD2, RTP801L, Smhs1
- Chromosome Location
- Chr. 3: 137623672 - 137628332 on Build GRCm38
- UniGene ID
- Mm.250841
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030143.4 |
NP_084419.2
|
2, 3 |
363 |
|
GenBank mRNA |
AF327512.1 |
- |
1, 2 |
118 |
|
AF335325.1 |
- |
1, 2 |
163 |
|
AK006617.1 |
- |
2, 3 |
362 |
|
AK018944.1 |
- |
2, 3 |
365 |
|
AK053333.1 |
- |
2, 3 |
301 |
|
AK075693.1 |
- |
2, 3 |
252 |
|
BC038131.1 |
- |
2, 3 |
262 |
|
BC147181.1 |
- |
1, 2 |
142 |
9056209 |
BC147182.1 |
- |
1, 2 |
142 |
9056220 |
View Full Product Details