Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026665.4 | NP_080941.3 | 9 | 1117 | |
GenBank mRNA | AK014862.1 | - | 6 | 750 | |
AK014873.1 | - | 9 | 1064 | ||
AK014982.1 | - | 7 | 575 | ||
AK016484.1 | - | 9 | 901 | ||
AK019414.1 | - | 9 | 997 | ||
AK028408.1 | - | 8 | 888 | ||
AK028458.1 | - | 9 | 945 | ||
AY225093.1 | - | 9 | 910 | ||
AY251192.1 | - | 9 | 1062 | ||
BC025588.1 | - | 8 | 897 | ||
BC050785.1 | - | 7 | 628 | 6703205 | |
BC056968.1 | - | 9 | 1361 |