Gene Transcripts

Gene Symbol
Lrrc17
Entrez Gene ID
74511
Gene Name
leucine rich repeat containing 17
Gene Aliases
37kDa, 4833425M04Rik, 6130400C22Rik, P37nb
Chromosome Location
Chr. 5: 21543527 - 21575911 on Build GRCm38
UniGene ID
Mm.439963
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028977.1 NP_083253.1 2 330
GenBank mRNA AK018071.1 - 2 296
BC030317.1 - 2 249 5372009
CF914556.1 - 2 330

View Full Product Details