Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029844.3 | NP_084120.1 | 1 | 285 | |
GenBank mRNA | AK003912.2 | - | 1 | 286 | |
AK158113.1 | - | 1 | 159 | ||
AY079153.1 | - | 1 | 62 | ||
AY079154.1 | - | 1 | 45 | ||
BC027543.1 | - | 1 | 139 | 1477097 |