Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152922.3 | NP_690886.3 | 6, 7 | 679 | |
GenBank mRNA | AK148723.1 | - | 6, 7 | 680 | |
AY635583.1 | - | 6, 7 | 680 | ||
BC055829.1 | - | 2, 3 | 1001 | ||
BC139484.1 | - | 6, 7 | 648 | 8862506 | |
BC141312.1 | - | 6, 7 | 648 | 9055924 | |
BC145632.1 | - | 6, 7 | 648 | 9054200 |