Product Details
- SNP ID
-
rs13481289
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:7977812 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGAAAGCCTGCCCTGCGGACGCCG[C/T]TACTGCTGCTGTTCCTGCTACTGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Apob
PubMed Links
Gene Details
- Gene
- Apob
- Gene Name
- apolipoprotein B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009693.2 |
136 |
Silent Mutation |
|
|
NP_033823.2 |
View Full Product Details