Product Details
- SNP ID
-
rs6401342
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:53391590 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCATAGATTTAAGCCTTTATGT[C/T]CTTTAATGTCTTCTCTCACTCACAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Npas3
PubMed Links
Gene Details
- Gene
- Npas3
- Gene Name
- neuronal PAS domain protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013780.2 |
|
Intron |
|
|
NP_038808.2 |
View Full Product Details