Product Details
- SNP ID
-
rs13481905
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:81107255 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCAGTCAGGTAAGAGGTCACTG[C/G]CAGTTCAAAGTCACTAAATGCTCAT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Adgrv1
PubMed Links
Gene Details
- Gene
- Adgrv1
- Gene Name
- adhesion G protein-coupled receptor V1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_054053.4 |
|
Intron |
|
|
NP_473394.3 |
View Full Product Details