Product Details
- SNP ID
-
rs13480660
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:77556536 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTCTCAGAGTGGGGAGACCCCA[A/G]AAATCAGCACAGTGGCCTTGGACCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Itgb2
PubMed Links
Gene Details
- Gene
- Itgb2
- Gene Name
- integrin beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008404.4 |
|
Intron |
|
|
NP_032430.2 |
View Full Product Details