Product Details
- SNP ID
-
rs13478694
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:32302244 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATTCATTCACCCCAAGGAAAAA[C/G]TCTCCATGGAGGGCAATGATGGCTG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Plxna4
PubMed Links
Gene Details
- Gene
- Plxna4
- Gene Name
- plexin A4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175750.3 |
|
Intron |
|
|
NP_786926.2 |
View Full Product Details