Product Details

SNP ID
rs13481246
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:115801702 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTGTGGACACATAAAGACATA[C/T]GGGGCAGGTCCTTGGCCAGGGAGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
2310067B10Rik PubMed Links

Gene Details

Gene
2310067B10Rik
Gene Name
RIKEN cDNA 2310067B10 gene
There are no transcripts associated with this gene.

Gene
Caskin2
Gene Name
CASK-interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080643.2 2737 Silent Mutation NP_542374.2
Gene
Mir5621
Gene Name
microRNA 5621
There are no transcripts associated with this gene.

View Full Product Details