Product Details

SNP ID
rs13481861
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:68572632 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTTTACGGCATCATTTGTAGTCAG[A/C]CGAATGGCTTTTGAAATTGGAACTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Fastkd3 PubMed Links

Gene Details

Gene
Fastkd3
Gene Name
FAST kinase domains 3
There are no transcripts associated with this gene.

Gene
Mtrr
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase reductase
There are no transcripts associated with this gene.

View Full Product Details