Product Details
- SNP ID
-
rs13477059
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:35788473 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAAGATGAAATTTTTCCTGCAGA[C/T]TTGGTGCTTCTGTCCTCAGATCGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Atp11b
PubMed Links
Gene Details
- Gene
- Atp11b
- Gene Name
- ATPase, class VI, type 11B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_029570.3 |
717 |
Silent Mutation |
|
|
NP_083846.2 |
View Full Product Details