Product Details

SNP ID
rs13481571
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:85943851 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAGTGGAACATAAAGCCATTC[G/T]ATTTCTTTCATTTCATTGCTCTTTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Ttll5 PubMed Links

Gene Details

Gene
Ttll5
Gene Name
tubulin tyrosine ligase-like family, member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081423.2 Intron NP_001074892.2

View Full Product Details