Product Details

SNP ID
rs3676875
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:52563134 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGTTGGGAAAAAAAAAAGAGTCAAA[T/C]TCTTGGGGGAAAAGTCCTATAATGC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Cacnb4 PubMed Links

Gene Details

Gene
Cacnb4
Gene Name
calcium channel, voltage-dependent, beta 4 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037099.2 Intron NP_001032176.1
NM_001285426.1 Intron NP_001272355.1
NM_001285427.1 Intron NP_001272356.1
NM_001285428.1 Intron NP_001272357.1
NM_146123.3 Intron NP_666235.1

View Full Product Details