Product Details
- SNP ID
-
rs33878164
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:60176343 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAAAACAGTGTATTTTTGAAAAG[A/T]GCAAAATAAGTCCCAAGCTGTACAC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
Mcf2
PubMed Links
Gene Details
- Gene
- Mcf2
- Gene Name
- mcf.2 transforming sequence
There are no transcripts associated with this gene.
View Full Product Details