Product Details
- SNP ID
-
rs13479671
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:28940533 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCACCAAGAGCATCAACAAAAAAG[A/G]TAAATGAAATTCAGAACAAAACTTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Unc5d
PubMed Links
Gene Details
- Gene
- Unc5d
- Gene Name
- unc-5 homolog D (C. elegans)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153135.3 |
|
Intron |
|
|
NP_694775.1 |
View Full Product Details