Product Details
- SNP ID
-
rs3022948
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:7443308 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGCTGTGTTTTGACATCACAATT[C/T]TTGTATGGGGAGTTTCTGTGGCCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pkia
PubMed Links
Gene Details
- Gene
- Pkia
- Gene Name
- protein kinase inhibitor, alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008862.3 |
1625 |
UTR 3 |
|
|
NP_032888.1 |
View Full Product Details