Product Details
- SNP ID
-
rs29140787
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:78603526 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCAGACCCTGTTGTTTCTGTCT[A/G]TCTAGTGTTTGTATGTTTGGATGGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Gphn
PubMed Links
Gene Details
- Gene
- Gphn
- Gene Name
- gephyrin
View Full Product Details