Product Details

SNP ID
rs13483049
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:57212916 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTGTTGAACACTTTTATGTG[G/T]TGTCTAGCTTAGTCTGTACATTAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C3 PubMed Links

Gene Details

Gene
C3
Gene Name
complement component 3
There are no transcripts associated with this gene.

Gene
Mir6978
Gene Name
microRNA 6978
There are no transcripts associated with this gene.

View Full Product Details