Product Details

SNP ID
rs6152313
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:76235050 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGTTCGCTCTCTGAAGCCCGA[A/C]AACCCACATTTGAAGAGATCACTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Pde11a PubMed Links

Gene Details

Gene
Pde11a
Gene Name
phosphodiesterase 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081033.1 Intron NP_001074502.1

View Full Product Details