Product Details
- SNP ID
-
rs13483334
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:44767434 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTATAAGAAGGGGGAAAACAGTA[C/G]AAAAACAGGCAAGTAATGTGACTAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Mcc
PubMed Links
Gene Details
- Gene
- Mcc
- Gene Name
- mutated in colorectal cancers
View Full Product Details