Product Details
- SNP ID
-
rs13480714
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:94047286 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGAAGAGTCCAGTGCAAACCCCA[A/T]AGGAGGGTCTGTTTGCTGAAAGTGG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Fgd6
PubMed Links
Gene Details
- Gene
- Fgd6
- Gene Name
- FYVE, RhoGEF and PH domain containing 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_053072.3 |
|
Intron |
|
|
NP_444302.4 |
View Full Product Details