Product Details

SNP ID
rs13481043
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:57484032 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGTTCTGTCAGAGGTCTCTGG[A/G]CTGTGGCTCACACGTGGATGACTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Fam114a2 PubMed Links

Gene Details

Gene
Fam114a2
Gene Name
family with sequence similarity 114, member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168667.1 1548 Missense Mutation NP_001162138.1
NM_001168668.1 1548 Missense Mutation NP_001162139.1
NM_026342.3 1548 Missense Mutation NP_080618.1

View Full Product Details